missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RPL10L Partial ORF (NP_542784.1, 115 a.a. - 214 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00140801-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MRGAFGKPQGTVARVHIGQVIMSIRTKLQNEEHVIEALRRAKFKFPGRQKIHISKKWGFTKFNADEFEDMVAKKCLIPDGCGVKYVPSHGPLDKWRVLHSEspecificaciones
NP_542784.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRGAFGKPQGTVARVHIGQVIMSIRTKLQNEEHVIEALRRAKFKFPGRQKIHISKKWGFTKFNADEFEDMVAKKCLIPDGCGVKYVPSHGPLDKWRVLHS | |
RUO | |
RPL10L | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
140801 | |
RPL10L (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ27353 | |
RPL10L | |
Yes |