missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human RP1-21O18.1 Full-length ORF (NP_001018000.1, 1 a.a. - 415 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023254-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MRAADSGSWERVRQLAAQGEPAPSCGAGAGPARPPGPAACEQCVDAAGPGDRPRAGVPRVRADGDCSQPVLLREEVSRLQEEVHLLRQMKEMLAKDLEESQGGKSSEVLSATELRVQLAQKEQELARAKEALQAMKADRKRLKGEKTDLVSQMQQLYATLESREEQLRDFIRNYEQHRKESEDAVKALAKEKDLLEREKWELRRQAKEATDHATALRSQLDLKDNRMKELEAELAMAKQSLATLTKDVPKRHSLAMPGETVLNGNQEWVVQADLPLTAAIRQSQQTLYHSHPPHPADRQAVRVSPCHSRQPSVISDASAAEGDRSSTPSDINSPRHRTHSLCNGDSPGPVQKNLHNPIVQSLEDLEDQKRKKKKEKMGFGSISRVFARGKQRKSLDPGLFDGTAPDYYIEEDADWEspecificaciones
NP_001018000.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
72.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686D08115/FLJ10199/FLJ32593/FLJ43398/FLJ43806/KIAA1026/MGC126686/MGC142181 | |
RP1-21O18.1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23254 | |
RP1-21O18.1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MRAADSGSWERVRQLAAQGEPAPSCGAGAGPARPPGPAACEQCVDAAGPGDRPRAGVPRVRADGDCSQPVLLREEVSRLQEEVHLLRQMKEMLAKDLEESQGGKSSEVLSATELRVQLAQKEQELARAKEALQAMKADRKRLKGEKTDLVSQMQQLYATLESREEQLRDFIRNYEQHRKESEDAVKALAKEKDLLEREKWELRRQAKEATDHATALRSQLDLKDNRMKELEAELAMAKQSLATLTKDVPKRHSLAMPGETVLNGNQEWVVQADLPLTAAIRQSQQTLYHSHPPHPADRQAVRVSPCHSRQPSVISDASAAEGDRSSTPSDINSPRHRTHSLCNGDSPGPVQKNLHNPIVQSLEDLEDQKRKKKKEKMGFGSISRVFARGKQRKSLDPGLFDGTAPDYYIEEDADW | |
RUO | |
RP1-21O18.1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |