Learn More
Abnova™ Human RNF36 Partial ORF (NP_542783, 33 a.a. - 121 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00140691-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a member of the RING-B-box-coiled-coil (RBCC) family and encodes a protein with an N-terminal RING finger motif, a PRY domain and a C-terminal SPRY domain. The mouse ortholog of this gene is specifically expressed in germ cells at the round spermatid stages during spermatogenesis and, when overexpressed, induces apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described. [provided by RefSeq]
Sequence: HKENKLHLQQHVSMEFLKLHQFLHSKEKDILTELREEGKALNEEMELNLSQLQEQCLLAKDMLVSIQAKTEQQNSFDFLKDITTLLHSLEspecificaciones
NP_542783 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.53kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HKENKLHLQQHVSMEFLKLHQFLHSKEKDILTELREEGKALNEEMELNLSQLQEQCLLAKDMLVSIQAKTEQQNSFDFLKDITTLLHSL | |
RUO | |
TRIM69 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
140691 | |
RNF36 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSD34/RNF36/Trif | |
TRIM69 | |
Recombinant | |
wheat germ expression system |