Learn More
Abnova™ Human RNF2 Full-length ORF (NP_009143.1, 1 a.a. - 336 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006045-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Polycomb group (PcG) of proteins form the multiprotein complexes that are important for the transcription repression of various genes involved in development and cell proliferation. The protein encoded by this gene is one of the PcG proteins. It has been shown to interact with, and suppress the activity of, transcription factor CP2 (TFCP2/CP2). Studies of the mouse counterpart suggested the involvement of this gene in the specification of anterior-posterior axis, as well as in cell proliferation in early development. This protein was also found to interact with huntingtin interacting protein 2 (HIP2), an ubiquitin-conjugating enzyme, and possess ubiquitin ligase activity. [provided by RefSeq]
Sequence: MSQAVQTNGTQPLSKTWELSLYELQRTPQEAITDGLEIVVSPRSLHSELMCPICLDMLKNTMTTKECLHRFCADCIITALRSGNKECPTCRKKLVSKRSLRPDPNFDALISKIYPSRDEYEAHQERVLARINKHNNQQALSHSIEEGLKIQAMNRLQRGKKQQIENGSGAEDNGDSSHCSNASTHSNQEAGPSNKRTKTSDDSGLELDNNNAAMAIDPVMDGASEIELVFRPHPTLMEKDDSAQTRYIKTSGNATVDHLSKYLAVRLALEELRSKGESNQMNLDTASEKQYTIYIATASGQFTVLNGSFSLELVSEKYWKVNKPMELYYAPTKEHKEspecificaciones
NP_009143.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BAP-1/BAP1/DING/HIPI3/RING1B/RING2 | |
RNF2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6045 | |
RNF2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSQAVQTNGTQPLSKTWELSLYELQRTPQEAITDGLEIVVSPRSLHSELMCPICLDMLKNTMTTKECLHRFCADCIITALRSGNKECPTCRKKLVSKRSLRPDPNFDALISKIYPSRDEYEAHQERVLARINKHNNQQALSHSIEEGLKIQAMNRLQRGKKQQIENGSGAEDNGDSSHCSNASTHSNQEAGPSNKRTKTSDDSGLELDNNNAAMAIDPVMDGASEIELVFRPHPTLMEKDDSAQTRYIKTSGNATVDHLSKYLAVRLALEELRSKGESNQMNLDTASEKQYTIYIATASGQFTVLNGSFSLELVSEKYWKVNKPMELYYAPTKEHK | |
RUO | |
RNF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |