missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF135 Full-length ORF (NP_922921.1, 1 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084282-P01.25ug

 View more versions of this product

Product Code. 16175773

  • 521.00€ / 25 microgramos

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore more special offers
This item is not returnable. View return policy

Description

Description

The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene is located in a chromosomal region known to be frequently deleted in patients with neurofibromatosis. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq]

Sequence: MAGLGLGSAVPVWLAEDDLGCIICQGLLDWPATLPCGHSFCRHCLEALWGARDARRWACPTCRQGAAQQPHLRKNTLLQDLADKYRRAAREIQAGSDPAHCPCPGSSSLSSAAARPRRRPELQRVAVEKSITEVAQELTELVEHLVDIVRSLQNQRPLSESGPDNELSILGKENSWKPRLPPHAHCLTRATLHSGELLGLLSGPSIQPLT
Specifications

Specifications

NP_922921.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.3kDa
Glutathione Sepharose 4 Fast Flow
25 μg
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
L13/MGC13061/Riplet
RNF135
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84282
RNF135 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGLGLGSAVPVWLAEDDLGCIICQGLLDWPATLPCGHSFCRHCLEALWGARDARRWACPTCRQGAAQQPHLRKNTLLQDLADKYRRAAREIQAGSDPAHCPCPGSSSLSSAAARPRRRPELQRVAVEKSITEVAQELTELVEHLVDIVRSLQNQRPLSESGPDNELSILGKENSWKPRLPPHAHCLTRATLHSGELLGLLSGPSIQPLT
RUO
RNF135
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers