missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RNF135 Full-length ORF (NP_922921.1, 1 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00084282-P01.10ug

 Ver más versiones de este producto

Código de producto. 16165773

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. This gene is located in a chromosomal region known to be frequently deleted in patients with neurofibromatosis. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq]

Sequence: MAGLGLGSAVPVWLAEDDLGCIICQGLLDWPATLPCGHSFCRHCLEALWGARDARRWACPTCRQGAAQQPHLRKNTLLQDLADKYRRAAREIQAGSDPAHCPCPGSSSLSSAAARPRRRPELQRVAVEKSITEVAQELTELVEHLVDIVRSLQNQRPLSESGPDNELSILGKENSWKPRLPPHAHCLTRATLHSGELLGLLSGPSIQPLT
Especificaciones

Especificaciones

NP_922921.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.3kDa
Glutathione Sepharose 4 Fast Flow
10 μg
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
L13/MGC13061/Riplet
RNF135
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84282
RNF135 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAGLGLGSAVPVWLAEDDLGCIICQGLLDWPATLPCGHSFCRHCLEALWGARDARRWACPTCRQGAAQQPHLRKNTLLQDLADKYRRAAREIQAGSDPAHCPCPGSSSLSSAAARPRRRPELQRVAVEKSITEVAQELTELVEHLVDIVRSLQNQRPLSESGPDNELSILGKENSWKPRLPPHAHCLTRATLHSGELLGLLSGPSIQPLT
RUO
RNF135
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales