missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RIOK3 Partial ORF (AAH39729, 411 a.a. - 516 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008780-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16199235

  • 458.00€ / 25 microgramos


Fecha estimada de envío
19-10-2020
Añadir a la cesta

Descripción

Descripción

Sequence: NMLWHAGKVWLIDVSQSVEPTHPHGLEFLFRDCRNVSQKGGVKEALSERELFNAVSGLNITADNEADFLAEIEALEKMNEDHVQKNGRKAASFLKDDGDPPLLYDE
Especificaciones

Especificaciones

AAH39729
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.29kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NMLWHAGKVWLIDVSQSVEPTHPHGLEFLFRDCRNVSQKGGVKEALSERELFNAVSGLNITADNEADFLAEIEALEKMNEDHVQKNGRKAASFLKDDGDPPLLYDE
RUO
RIOK3
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
8780
RIOK3 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp779L1370|SUDD
RIOK3
Yes
Videos
Documentos