missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RGC32 Partial ORF (NP_054778.1, 1 a.a. - 117 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00028984-Q02.10ug

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 16015455

  • 349.00€ / 10 microgramos
Fecha estimada de envío: 08-11-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is thought to regulate cell cycle progression. It is induced by p53 in response to DNA damage, or by sublytic levels of complement system proteins that result in activation of the cell cycle. The encoded protein localizes to the cytoplasm during interphase and to centrosomes during mitosis. The protein forms a complex with polo-like kinase 1. The protein also translocates to the nucleus in response to treatment with complement system proteins, and can associate with and increase the kinase activity of cell division cycle 2 protein. In different assays and cell types, overexpression of this protein has been shown to activate or suppress cell cycle progression. [provided by RefSeq]

Sequence: MKPPAEDLSDALCEFDAVLADFASPFHERHFHYEEHLERMKRRSSASVSDSSGFSDSESADSLYRNSFSFSDEKLNSPTDSTPALLSATVTPQKAKLGDTKELEAFIADLDKTLASM
Especificaciones

Especificaciones

NP_054778.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.61kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MKPPAEDLSDALCEFDAVLADFASPFHERHFHYEEHLERMKRRSSASVSDSSGFSDSESADSLYRNSFSFSDEKLNSPTDSTPALLSATVTPQKAKLGDTKELEAFIADLDKTLASM
RUO
C13orf15
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
28984
RGC32 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0564/MGC87338/RGC-32/RGC32/bA157L14.2
C13orf15
Recombinant
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales