Learn More
Abnova™ Human REG1A Full-length ORF (NP_002900.2, 1 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005967-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is a type I subclass member of the Reg gene family. The Reg gene family is a multigene family grouped into four subclasses, types I, II, III and IV, based on the primary structures of the encoded proteins. This gene encodes a protein that is secreted by the exocrine pancreas. It is associated with islet cell regeneration and diabetogenesis and may be involved in pancreatic lithogenesis. Reg family members REG1B, REGL, PAP and this gene are tandemly clustered on chromosome 2p12 and may have arisen from the same ancestral gene by gene duplication. [provided by RefSeq]
Sequence: MAQTSSYFMLISCLMFLSQSQGQEAQTELPQARISCPEGTNAYRSYCYYFNEDRETWVDADLYCQNMNSGNLVSVLTQAEGAFVASLIKESGTDDFNVWIGLHDPKKNRRWHWSSGSLVSYKSWGIGAPSSVNPGYCVSLTSSTGFQKWKDVPCEDKFSFVCKFKNEspecificaciones
NP_002900.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ICRF/MGC12447/P19/PSP/PSPS/PSPS1/PTP/REG | |
REG1A | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5967 | |
REG1A (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MAQTSSYFMLISCLMFLSQSQGQEAQTELPQARISCPEGTNAYRSYCYYFNEDRETWVDADLYCQNMNSGNLVSVLTQAEGAFVASLIKESGTDDFNVWIGLHDPKKNRRWHWSSGSLVSYKSWGIGAPSSVNPGYCVSLTSSTGFQKWKDVPCEDKFSFVCKFKN | |
RUO | |
REG1A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |