Learn More
STEMCELL Technologies Human Recombinant TGF-beta 3
Transforming growth factor beta 3
Marca:  STEMCELL Technologies 78131.2
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17109641
  • 6212.43€ / paquete de 4
Fecha estimada de envío: 22-04-2024
para ver el stock
Descripción
Descripción
  • Transforming growth factor (TGF) beta 3 is a member of the TGF-β superfamily and regulates diverse cellular phenotypes
  • TGF-beta 3 binds to serine-threonine kinase type I and II receptors and activates signal transduction through SMAD2/3 proteins, thus regulating a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, and metabolism (Massagué; McDowell et al.) TGF-beta 3 enhances glycosaminoglycan production by mesenchymal stromal cells, stimulates scar-free healing, and improves glucose tolerance and phenotypic changes in adipocyte morphology (Hall et al.; Holton et al.) TGF-beta 3 induces proliferation of posterofrontal suture-derived mesenchymal stromal cells, and stimulates expression of fibroblast growth factors 2 and 18 (James et al.)
Especificaciones
Especificaciones
ARVD, FLJ16571, TGFB3, TGF-b3, Transforming growth factor beta-3
4 x 250 μg
Mesenchymal Cells, PSC-Derived; Mesenchymal Stem and Progenitor Cells
≥95%