Learn More
STEMCELL Technologies Human Recombinant G-CSF, ACF
Granulocyte colony-stimulating factor, animal component-free
Marca: STEMCELL Technologies 78138.1
Detalles adicionales : Peso : 0.00110kg
Descripción
- Granulocyte colony-stimulating factor (G-CSF) is a member of the CSF family of glycoproteins that regulate hematopoietic cell proliferation, differentiation, and function
- It is a key cytokine involved in the production of neutrophils and the stimulation of granulocyte colony formation from hematopoietic progenitor cells
- G-CSF causes a range of effects including a transient reduction of SDF-1 expression, the activation of metalloproteases that cleave VCAM-1, and the release of norepinephrine from the sympathetic nervous system, leading to the release or mobilization of hematopoietic stem cells from the bone marrow into the periphery
- The G-CSF receptor is expressed on a variety of hematopoietic cells, including myeloid-committed progenitor cells, neutrophils, granulocytes, and monocytes
Especificaciones
Colony-stimulating factor 3, CSF-3, MGI-1G, Pluripoietin | |
100 μg | |
Cancer Cells and Cell Lines; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neurons | |
≥95% |