Learn More
STEMCELL Technologies Human Recombinant EGF
Epidermal growth factor
Marca:  STEMCELL Technologies 78006.2
Detalles adicionales : Peso : 0.00110kg
 Ver más versiones de este producto
Código de producto. 17119331
  • 308.56€ / 1000 microgramos
Fecha estimada de envío: 04-04-2024
para ver el stock
Descripción
Descripción
  • Epidermal growth factor (EGF) is characterized by high affinity binding to various EGF receptors (EGFRs) and the production of mitogenic responses (Carpenter & Cohen)
  • EGF promotes EGFR dimerization, resulting in activation of downstream pathways including PI3K, ERK1/2, JAK/STAT, β-catenin, and calcium signaling
  • EGF is secreted by the gut-associated salivary and Brunner's glands, is found in a variety of body fluids, and stimulates cell proliferation and differentiation in rodent and neonatal human intestine (Wright et al.) Central nervous system stem cells also proliferate in response to the EGF stimulus (Reynolds & Weiss)
Especificaciones
Especificaciones
Beta-urogastrone, HOMG4, Epidermal growth factor, Pro-epidermal growth factor, URG, Urogastrone
1000 μg
Brain Tumor Stem Cells; Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Pluripotent Stem Cells; Prostate Cells
>95%