missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human RBM24 Full-length ORF (BAB70914.1, 1 a.a. - 191 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00221662-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16199483

  • 302.00€ / 10 microgramos


Fecha estimada de envío
19-10-2020
Añadir a la cesta

Descripción

Descripción

Sequence: MYVCLCVSVAKVTMADRAAAERACKDPNPIIDGRKANVNLAYLGAKPRIMQPGFAFGVQQLHPALIQRPFGIPAHYVYPQAFVQPGVVIPHVQPTAAAASTTPYIDYTGAAYAQYSAAAAAAAAAAAYDQYPYAASPAAAGYVTAGGYGYAVQQPITAAAPGTAAAAAAAAAAAAAFGQYQPQQLQTDRMQ
Especificaciones

Especificaciones

BAB70914.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ26355|FLJ30829|FLJ37697|RNPC6|dJ259A10.1
RBM24
Yes
wheat germ expression system
Liquid
221662
RBM24 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MYVCLCVSVAKVTMADRAAAERACKDPNPIIDGRKANVNLAYLGAKPRIMQPGFAFGVQQLHPALIQRPFGIPAHYVYPQAFVQPGVVIPHVQPTAAAASTTPYIDYTGAAYAQYSAAAAAAAAAAAYDQYPYAASPAAAGYVTAGGYGYAVQQPITAAAPGTAAAAAAAAAAAAAFGQYQPQQLQTDRMQ
RUO
RBM24
Wheat Germ (in vitro)
GST
Videos
Documentos