Learn More
Abnova™ Human RBBP5 Partial ORF (NP_005048.2, 392 a.a. - 491 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005929-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that bind directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B. [provided by RefSeq]
Sequence: ELEDSKALLYLPIAPEVEDPEENPYGPPPDAVQTSLMDEGASSEKKRQSSADGSQPPKKKPKTTNIELQGVPNDEVHPLLGVKGDGKSKKKQAGRPKGSKEspecificaciones
NP_005048.2 | |
Liquid | |
5929 | |
RBBP5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RBQ3/SWD1 | |
RBBP5 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ELEDSKALLYLPIAPEVEDPEENPYGPPPDAVQTSLMDEGASSEKKRQSSADGSQPPKKKPKTTNIELQGVPNDEVHPLLGVKGDGKSKKKQAGRPKGSK | |
RUO | |
RBBP5 | |
Wheat Germ (in vitro) | |
GST |