missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PTK9 Full-length ORF (NP_002813.2, 1 a.a. - 384 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005756-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16199881

  • 302.00€ / 10 microgramos


Fecha estimada de envío
26-10-2020
Añadir a la cesta

Descripción

Descripción

Sequence: MTRRGRRRSSHFLGPPAGAAGCTQRRSRELAAAAMSHQTGIQASEDVKEIFARARNGKYRLLKISIENEQLVIGSYSQPSDSWDKDYDSFVLPLLEDKQPCYILFRLDSQNAQGYEWIFIAWSPDHSHVRQKMLYAATRATLKKEFGGGHIKDEVFGTVKEDVSLHGYKKYLLSQSSPAPLTAAEEELRQIKINEVQTDVGVDTKHQTLQGVAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSFAKPKGPAGKRGIRRLIRGPAETEATTD
Especificaciones

Especificaciones

NP_002813.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
A6|MGC23788|MGC41876|PTK9
TWF1
Yes
wheat germ expression system
Liquid
5756
PTK9 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTRRGRRRSSHFLGPPAGAAGCTQRRSRELAAAAMSHQTGIQASEDVKEIFARARNGKYRLLKISIENEQLVIGSYSQPSDSWDKDYDSFVLPLLEDKQPCYILFRLDSQNAQGYEWIFIAWSPDHSHVRQKMLYAATRATLKKEFGGGHIKDEVFGTVKEDVSLHGYKKYLLSQSSPAPLTAAEEELRQIKINEVQTDVGVDTKHQTLQGVAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSFAKPKGPAGKRGIRRLIRGPAETEATTD
RUO
TWF1
Wheat Germ (in vitro)
GST
Videos
Documentos