missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PRMT7 Full-length ORF (NP_061896.1, 1 a.a. - 692 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00054496-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16100583

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Arginine methylation is an apparently irreversible protein modification catalyzed by arginine methyltransferases, such as PMT7, using S-adenosylmethionine (AdoMet) as the methyl donor. Arginine methylation is implicated in signal transduction, RNA transport, and RNA splicing (Miranda et al., 2004 [PubMed 15044439]).[supplied by OMIM]

Sequence: MKIFCSRANPTTGSVEWLEEDEHYDYHQEIARSSYADMLHDKDRNVKYYQGIRAAVSRVKDRGQKALVLDIGTGTGLLSMMAVTAGADFCYAIEVFKPMADAAVKIVEKNGFSDKIKVINKHSTEVTVGPEGDMPCRANILVTELFDTELIGEGALPSYEHAHRHLVEENCEAVPHRATVYAQLVESGRMWSWNKLFPIHVQTSLGEQVIVPPVDVESCPGAPSVCDIQLNQVSPADFTVLSDVLPMFSIDFSKQVSSSAACHSRRFEPLTSGRAQVVLSWWDIEMDPEGKIKCTMAPFWAHSDPEEMQWRDHWMQCVYFLPQEEPVVQGSALYLVAHHDDYCVWYSLQRTSPEKNERVRQMRPVCDCQAHLLWNRPRFGEINDQDRTDRYVQALRTVLKPDSVCLCVSDGSLLSVLAHHLGVEQVFTVESSAASHKLLRKIFKANHLEDKINIIEKRPELLTNEDLQGRKVSLLLGEPFFTTSLLPWHNLYFWYVRTAVDQHLGPGAMVMPQAASLHAVVVEFRDLWRIRSPCGDCEGFDVHIMDDMIKRALDFRESREAEPHPLWEYPCRSLSEPWQILTFDFQQPVPLQPLCAEGTVELRRPGQSHAAVLWMEYHLTPECTLSTGLLEPADPEGGCCWNPHCKQAVYFFSPAPDPRALLGGPRTVSYAVEFHPDTGDIIMEFRHADTPD
Especificaciones

Especificaciones

NP_061896.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
104.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ10640/KIAA1933
PRMT7
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
54496
PRMT7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKIFCSRANPTTGSVEWLEEDEHYDYHQEIARSSYADMLHDKDRNVKYYQGIRAAVSRVKDRGQKALVLDIGTGTGLLSMMAVTAGADFCYAIEVFKPMADAAVKIVEKNGFSDKIKVINKHSTEVTVGPEGDMPCRANILVTELFDTELIGEGALPSYEHAHRHLVEENCEAVPHRATVYAQLVESGRMWSWNKLFPIHVQTSLGEQVIVPPVDVESCPGAPSVCDIQLNQVSPADFTVLSDVLPMFSIDFSKQVSSSAACHSRRFEPLTSGRAQVVLSWWDIEMDPEGKIKCTMAPFWAHSDPEEMQWRDHWMQCVYFLPQEEPVVQGSALYLVAHHDDYCVWYSLQRTSPEKNERVRQMRPVCDCQAHLLWNRPRFGEINDQDRTDRYVQALRTVLKPDSVCLCVSDGSLLSVLAHHLGVEQVFTVESSAASHKLLRKIFKANHLEDKINIIEKRPELLTNEDLQGRKVSLLLGEPFFTTSLLPWHNLYFWYVRTAVDQHLGPGAMVMPQAASLHAVVVEFRDLWRIRSPCGDCEGFDVHIMDDMIKRALDFRESREAEPHPLWEYPCRSLSEPWQILTFDFQQPVPLQPLCAEGTVELRRPGQSHAAVLWMEYHLTPECTLSTGLLEPADPEGGCCWNPHCKQAVYFFSPAPDPRALLGGPRTVSYAVEFHPDTGDIIMEFRHADTPD
RUO
PRMT7
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales