missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPP4R1 Partial ORF (NP_005125, 834 a.a. - 932 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00009989-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16191576

  • 467.00€ / 25 microgramos


Fecha estimada de envío
16-02-2021
Añadir a la cesta

Descripción

Descripción

Sequence: VIEDDCLPMDQFAVHLMPHLLTLANDRVPNVRVLLAKTLRQTLLEKDYFLASASCHQEAVEQTIMALQMDRDSDVKYFASIHPASTKISEDAMSTASST
Especificaciones

Especificaciones

NP_005125
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VIEDDCLPMDQFAVHLMPHLLTLANDRVPNVRVLLAKTLRQTLLEKDYFLASASCHQEAVEQTIMALQMDRDSDVKYFASIHPASTKISEDAMSTASST
RUO
PPP4R1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
9989
PPP4R1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PP4(Rmeg)|PP4R1
PPP4R1
Yes
Videos
SDS
Documentos