missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PPP2R5A Full-length ORF (NP_006234.1, 1 a.a. - 486 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005525-P02.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MSSSSPPAGAASAAISASEKVDGFTRKSVRKAQRQKRSQGSSQFRSQGSQAELHPLPQLKDATSNEQQELFCQKLQQCCILFDFMDSVSDLKSKEIKRATLNELVEYVSTNRGVIVESAYSDIVKMISANIFRTLPPSDNPDFDPEEDEPTLEASWPHIQLVYEFFLRFLESPDFQPSIAKRYIDQKFVQQLLELFDSEDPRERDFLKTVLHRIYGKFLGLRAFIRKQINNIFLRFIYETEHFNGVAELLEILGSIINGFALPLKAEHKQFLMKVLIPMHTAKGLALFHAQLAYCVVQFLEKDTTLTEPVIRGLLKFWPKTCSQKEVMFLGEIEEILDVIEPTQFKKIEEPLFKQISKCVSSSHFQVAERALYFWNNEYILSLIEENIDKILPIMFASLYKISKEHWNPTIVALVYNVLKTLMEMNGKLFDDLTSSYKAERQREKKKELEREELWKKLEELKLKKALEKQNSAYNMHSILSNTSAEEspecificaciones
NP_006234.1 | |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B56A|MGC131915|PR61A | |
PPP2R5A | |
Yes |
wheat germ expression system | |
Liquid | |
5525 | |
PPP2R5A (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSSSSPPAGAASAAISASEKVDGFTRKSVRKAQRQKRSQGSSQFRSQGSQAELHPLPQLKDATSNEQQELFCQKLQQCCILFDFMDSVSDLKSKEIKRATLNELVEYVSTNRGVIVESAYSDIVKMISANIFRTLPPSDNPDFDPEEDEPTLEASWPHIQLVYEFFLRFLESPDFQPSIAKRYIDQKFVQQLLELFDSEDPRERDFLKTVLHRIYGKFLGLRAFIRKQINNIFLRFIYETEHFNGVAELLEILGSIINGFALPLKAEHKQFLMKVLIPMHTAKGLALFHAQLAYCVVQFLEKDTTLTEPVIRGLLKFWPKTCSQKEVMFLGEIEEILDVIEPTQFKKIEEPLFKQISKCVSSSHFQVAERALYFWNNEYILSLIEENIDKILPIMFASLYKISKEHWNPTIVALVYNVLKTLMEMNGKLFDDLTSSYKAERQREKKKELEREELWKKLEELKLKKALEKQNSAYNMHSILSNTSAE | |
RUO | |
PPP2R5A | |
Wheat Germ (in vitro) | |
GST |