missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PP Partial ORF (NP_066952, 10 a.a. - 111 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005464-Q01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16199371

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme. [provided by RefSeq]

Sequence: AAPFSLEYRVFLKNEKGQYISPFHDIPIYADKDVFHMVVEVPRWSNAKMEIATKDPLNPIKQDVKKGKLRYVANLFPYKGYIWNYGAIPQTWEDPGHNDKHT
Especificaciones

Especificaciones

NP_066952
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.96kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AAPFSLEYRVFLKNEKGQYISPFHDIPIYADKDVFHMVVEVPRWSNAKMEIATKDPLNPIKQDVKKGKLRYVANLFPYKGYIWNYGAIPQTWEDPGHNDKHT
RUO
PPA1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5464
PP (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IOPPP/MGC111556/PP/PP1/SID6-8061
PPA1
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales