Learn More
Abcam Human PLSCR1 (Scramblase 1) knockout A549 cell line
Product Code. 32157991 Shop All Abcam Products
Change view
Click to view available options
Quantity:
2 x 1,000,000 Cells
Unit Size:
Pack of 2
Product Code. Quantity Unit Size
32157991 2 x 1,000,000 Cells Pack of 2
1 options
This item is not returnable. View return policy
Product Code. 32157991 Supplier Abcam Supplier No. ab2669882x1000000Cells_
 Restricted Item
Fecha estimada de envoi: 07-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
PLSCR1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 4 and 1 bp insertion in exon 4. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knocko...
PLSCR1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 4 and 1 bp insertion in exon 4. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells
Regulatory Status RUO