Learn More
Abnova™ Human PLEKHG5 Partial ORF (NP_065682, 896 a.a. - 995 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00057449-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that activates the nuclear factor kappa B (NFKB1) signaling pathway. Multations in this gene have been found in a family with distal spinal muscular atrophy. [provided by RefSeq]
Sequence: SAPSRSLSELCLAVPAPGIRTQGSPQEAGPSWDCRGAPSPGSGPGLVGCLAGEPAGSHRKRCGDLPSGASPRVQPEPPPGVSAQHRKLTLAQLYRIRTTLEspecificaciones
NP_065682 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SAPSRSLSELCLAVPAPGIRTQGSPQEAGPSWDCRGAPSPGSGPGLVGCLAGEPAGSHRKRCGDLPSGASPRVQPEPPPGVSAQHRKLTLAQLYRIRTTL | |
RUO | |
PLEKHG5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57449 | |
PLEKHG5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DSMA4/GEF720/KIAA0720/RP4-650H14.3 | |
PLEKHG5 | |
Recombinant | |
wheat germ expression system |