Learn More
Abnova™ Human PLA2G12A Full-length ORF (NP_110448.2, 1 a.a. - 189 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00081579-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Secreted phospholipase A2 (sPLA2) enzymes liberate arachidonic acid from phospholipids for production of eicosanoids and exert a variety of physiologic and pathologic effects. Group XII sPLA2s, such as PLA2G12A, have relatively low specific activity and are structurally and functionally distinct from other sPLA2s (Gelb et al., 2000 [PubMed 11031251]).[supplied by OMIM]
Sequence: MALLSRPALTLLLLLMAAVVRCQEQAQTTDWRATLKTIRNGVHKIDTYLNAALDLLGGEDGLCQYKCSDGSKPFPRYGYKPSPPNGCGSPLFGVHLNIGIPSLTKCCNQHDRCYETCGKSKNDCDEEFQYCLSKICRDVQKTLGLTQHVQACETTVELLFDSVIHLGCKPYLDSQRAACRCHYEEKTDLEspecificaciones
NP_110448.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
47.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GXII/PLA2G12/ROSSY | |
PLA2G12A | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
81579 | |
PLA2G12A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALLSRPALTLLLLLMAAVVRCQEQAQTTDWRATLKTIRNGVHKIDTYLNAALDLLGGEDGLCQYKCSDGSKPFPRYGYKPSPPNGCGSPLFGVHLNIGIPSLTKCCNQHDRCYETCGKSKNDCDEEFQYCLSKICRDVQKTLGLTQHVQACETTVELLFDSVIHLGCKPYLDSQRAACRCHYEEKTDL | |
RUO | |
PLA2G12A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |