Learn More
Abnova™ Human PKP4 Partial ORF (NP_001005476, 12 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Especificaciones
Número de acceso | NP_001005476 |
---|---|
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 8502 |
Peso molecular | 36.63kDa |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16198655
|
Abnova™
H00008502-Q01.25UG |
25 ug |
508.00€
25 microgramos |
Fecha estimada de envío: 30-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16188655
|
Abnova™
H00008502-Q01.10UG |
10 ug |
335.00€
10 microgramos |
Fecha estimada de envío: 30-05-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Armadillo-like proteins are characterized by a series of armadillo repeats, first defined in the Drosophila 'armadillo' gene product, that are typically 42 to 45 amino acids in length. These proteins can be divided into subfamilies based on their number of repeats, their overall sequence similarity, and the dispersion of the repeats throughout their sequences. Members of the p120(ctn)/plakophilin subfamily of Armadillo-like proteins, including CTNND1, CTNND2, PKP1, PKP2, PKP4, and ARVCF. PKP4 may be a component of desmosomal plaque and other adhesion plaques and is thought to be involved in regulating junctional plaque organization and cadherin function. Multiple transcript variants have been found for this gene, but the full-length nature of only two of them have been described so far. These two variants encode distinct isoforms. [provided by RefSeq]
Sequence: EGQPQTRQEAASTGPGMEPETTATTILASVKEQELQFQRLTRELEVERQIVASQLERCRLGAESPSIASTSSTEKSFPWRSTDVPNTGVSKPRVSDAVQEspecificaciones
NP_001005476 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ31261/FLJ42243/p0071 | |
PKP4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8502 | |
PKP4 (Human) Recombinant Protein (Q01) | |
EGQPQTRQEAASTGPGMEPETTATTILASVKEQELQFQRLTRELEVERQIVASQLERCRLGAESPSIASTSSTEKSFPWRSTDVPNTGVSKPRVSDAVQ | |
RUO | |
PKP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |