Learn More
Abnova™ Human PITPNB Partial ORF (NP_036531, 181 a.a. - 271 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00023760-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is found in the cytoplasm, where it catalyzes the transfer of phosphatidylinositol and phosphatidylcholine between membranes. [provided by RefSeq]
Sequence: LANSPDCPQMCAYKLVTIKFKWWGLQSKVENFIQKQEKRIFTNFHRQLFCWIDKWIDLTMEDIRRMEDETQKELETMRKRGSVRGTSAADVEspecificaciones
NP_036531 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LANSPDCPQMCAYKLVTIKFKWWGLQSKVENFIQKQEKRIFTNFHRQLFCWIDKWIDLTMEDIRRMEDETQKELETMRKRGSVRGTSAADV | |
RUO | |
PITPNB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
23760 | |
PITPNB (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PI-TP-beta/PtdInsTP/VIB1B | |
PITPNB | |
Recombinant | |
wheat germ expression system |