Learn More
Abnova™ Human PHIP Partial ORF (NP_060404, 1599 a.a. - 1705 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00055023-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
PHIP binds the pleckstrin homology (PH) domain of insulin receptor substrate-1 (IRS1; MIM 147545), modulates insulin signaling, and plays a role in pancreatic beta cell growth and survival (Farhang-Fallah et al., 2000 [PubMed 11018022]; Podcheko et al., 2007 [PubMed 17636024]).[supplied by OMIM]
Sequence: TLSKSSAVIEQGDCKNNALVPGTIQVNGHGGQPSKLVKRGPGRKPKVEVNTNSGEIIHKKRGRKPKKLQYAKPEDLEQNNVHPIRDEVLPSSTCNFLSETNNVKEDLEspecificaciones
NP_060404 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.51kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TLSKSSAVIEQGDCKNNALVPGTIQVNGHGGQPSKLVKRGPGRKPKVEVNTNSGEIIHKKRGRKPKKLQYAKPEDLEQNNVHPIRDEVLPSSTCNFLSETNNVKEDL | |
RUO | |
PHIP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55023 | |
PHIP (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ20705/FLJ45918/MGC90216/WDR11/ndrp | |
PHIP | |
Recombinant | |
wheat germ expression system |