Learn More
Abnova™ Human PGBD2 Full-length ORF (NP_733843.1, 1 a.a. - 592 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00267002-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. The exact function of this gene is not known. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MASTSRDVIAGRGIHSKVKSAKLLEVLNAMEEEESNNNREEIFIAPPDNAAGEFTDEDSGDEDSQRGAHLPGSVLHASVLCEDSGTGEDNDDLELQPAKKRQKAVVKPQRIWTKRDIRPDFGSWTASDPHIEDLKSQELSPVGLFELFFDEGTINFIVNETNRYAWQKNVNLSLTAQELKCVLGILILSGYISYPRRRMFWETSPDSHHHLVADAIRRDRFELIFSYLHFADNNELDASDRFAKVRPLIIRMNCNFQKHAPLEEFYSFGESMCEYFGHRGSKQLHRGKPVRLGYKIWCGTTSRGYLVWFEPSQGTLFTKPDRSLDLGGSMVIKFVDALQERGFLPYHIFFDKVFTSVKLMSILRKKGVKATGTVREYRTERCPLKDPKELKKMKRGSFDYKVDESEEIIVCRWHDSSVVNICSNAVGIEPVRLTSRHSGAAKTRTQVHQPSLVKLYQEKVGGVGRMDQNIAKYKVKIRGMKWYSSFIGYVIDAALNNAWQLHRICCQDAQVDLLAFRRYIACVYLESNADTTSQGRRSRRLETESRFDMIGHWIIHQDKRTRCALCHSQTNTRCEKCQKGVHAKCFREYHIREspecificaciones
NP_733843.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
94.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PGBD2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
267002 | |
PGBD2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASTSRDVIAGRGIHSKVKSAKLLEVLNAMEEEESNNNREEIFIAPPDNAAGEFTDEDSGDEDSQRGAHLPGSVLHASVLCEDSGTGEDNDDLELQPAKKRQKAVVKPQRIWTKRDIRPDFGSWTASDPHIEDLKSQELSPVGLFELFFDEGTINFIVNETNRYAWQKNVNLSLTAQELKCVLGILILSGYISYPRRRMFWETSPDSHHHLVADAIRRDRFELIFSYLHFADNNELDASDRFAKVRPLIIRMNCNFQKHAPLEEFYSFGESMCEYFGHRGSKQLHRGKPVRLGYKIWCGTTSRGYLVWFEPSQGTLFTKPDRSLDLGGSMVIKFVDALQERGFLPYHIFFDKVFTSVKLMSILRKKGVKATGTVREYRTERCPLKDPKELKKMKRGSFDYKVDESEEIIVCRWHDSSVVNICSNAVGIEPVRLTSRHSGAAKTRTQVHQPSLVKLYQEKVGGVGRMDQNIAKYKVKIRGMKWYSSFIGYVIDAALNNAWQLHRICCQDAQVDLLAFRRYIACVYLESNADTTSQGRRSRRLETESRFDMIGHWIIHQDKRTRCALCHSQTNTRCEKCQKGVHAKCFREYHIR | |
RUO | |
PGBD2 | |
Recombinant | |
wheat germ expression system |