missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PGBD1 Full-length ORF (NP_115896.1, 1 a.a. - 809 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00084547-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16105953

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The piggyBac family of proteins, found in diverse animals, are transposases related to the transposase of the canonical piggyBac transposon from the moth, Trichoplusia ni. This family also includes genes in several genomes, including human, that appear to have been derived from the piggyBac transposons. This gene belongs to the subfamily of piggyBac transposable element derived (PGBD) genes. The PGBD proteins appear to be novel, with no obvious relationship to other transposases, or other known protein families. This gene product is specifically expressed in the brain, however, its exact function is not known. [provided by RefSeq]

Sequence: MYEALPGPAPENEDGLVKVKEEDPTWEQVCNSQEGSSHTQEICRLRFRHFCYQEAHGPQEALAQLRELCHQWLRPEMHTKEQIMELLVLEQFLTILPKELQPCVKTYPLESGEEAVTVLENLETGSGDTGQQASVYIQGQDMHPMVAEYQGVSLECQSLQLLPGITTLKCEPPQRPQGNPQEVSGPVPHGSAHLQEKNPRDKAVVPVFNPVRSQTLVKTEEETAQAVAAEKWSHLSLTRRNLCGNSAQETVMSLSPMTEEIVTKDRLFKAKQETSEEMEQSGEASGKPNRECAPQIPCSTPIATERTVAHLNTLKDRHPGDLWARMHISSLEYAAGDITRKGRKKDKARVSELLQGLSFSGDSDVEKDNEPEIQPAQKKLKVSCFPEKSWTKRDIKPNFPSWSALDSGLLNLKSEKLNPVELFELFFDDETFNLIVNETNNYASQKNVSLEVTVQEMRCVFGVLLLSGFMRHPRREMYWEVSDTDQNLVRDAIRRDRFELIFSNLHFADNGHLDQKDKFTKLRPLIKQMNKNFLLYAPLEEYYCFDKSMCECFDSDQFLNGKPIRIGYKIWCGTTTQGYLVWFEPYQEESTMKVDEDPDLGLGGNLVMNFADVLLERGQYPYHLCFDSFFTSVKLLSALKKKGVRATGTIRENRTEKCPLMNVEHMKKMKRGYFDFRIEENNEIILCRWYGDGIISLCSNAVGIEPVNEVSCCDADNEEIPQISQPSIVKVYDECKEGVAKMDQIISKYRVRIRSKKWYSILVSYMIDVAMNNAWQLHRACNPGASLDPLDFRRFVAHFYLEHNAHLSD
Especificaciones

Especificaciones

NP_115896.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
118.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HUCEP-4/SCAND4/dJ874C20.4
PGBD1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84547
PGBD1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MYEALPGPAPENEDGLVKVKEEDPTWEQVCNSQEGSSHTQEICRLRFRHFCYQEAHGPQEALAQLRELCHQWLRPEMHTKEQIMELLVLEQFLTILPKELQPCVKTYPLESGEEAVTVLENLETGSGDTGQQASVYIQGQDMHPMVAEYQGVSLECQSLQLLPGITTLKCEPPQRPQGNPQEVSGPVPHGSAHLQEKNPRDKAVVPVFNPVRSQTLVKTEEETAQAVAAEKWSHLSLTRRNLCGNSAQETVMSLSPMTEEIVTKDRLFKAKQETSEEMEQSGEASGKPNRECAPQIPCSTPIATERTVAHLNTLKDRHPGDLWARMHISSLEYAAGDITRKGRKKDKARVSELLQGLSFSGDSDVEKDNEPEIQPAQKKLKVSCFPEKSWTKRDIKPNFPSWSALDSGLLNLKSEKLNPVELFELFFDDETFNLIVNETNNYASQKNVSLEVTVQEMRCVFGVLLLSGFMRHPRREMYWEVSDTDQNLVRDAIRRDRFELIFSNLHFADNGHLDQKDKFTKLRPLIKQMNKNFLLYAPLEEYYCFDKSMCECFDSDQFLNGKPIRIGYKIWCGTTTQGYLVWFEPYQEESTMKVDEDPDLGLGGNLVMNFADVLLERGQYPYHLCFDSFFTSVKLLSALKKKGVRATGTIRENRTEKCPLMNVEHMKKMKRGYFDFRIEENNEIILCRWYGDGIISLCSNAVGIEPVNEVSCCDADNEEIPQISQPSIVKVYDECKEGVAKMDQIISKYRVRIRSKKWYSILVSYMIDVAMNNAWQLHRACNPGASLDPLDFRRFVAHFYLEHNAHLSD
RUO
PGBD1
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales