missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PEX13 Full-length ORF (NP_002609.1, 1 a.a. - 403 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005194-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16118941

  • 528.00€ / 25 microgramos
Fecha estimada de envío: 17-07-2023
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a peroxisomal membrane protein that binds the type 1 peroxisomal targeting signal receptor via a SH3 domain located in the cytoplasm. Mutations and deficiencies in peroxisomal protein importing and peroxisome assembly lead to peroxisomal biogenesis disorders, an example of which is Zellweger syndrome. [provided by RefSeq]

Sequence: MASQPPPPPKPWETRRIPGAGPGPGPGPTFQSADLGPTLMTRPGQPALTRVPPPILPRPSQQTGSSSVNTFRPAYSSFSSGYGAYGNSFYGGYSPYSYGYNGLGYNRLRVDDLPPSRFVQQAEESSRGAFQSIESIVHAFASVSMMMDATFSAVYNSFRAVLDVANHFSRLKIHFTKVFSAFALVRTIRYLYRRLQRMLGLRRGSENEDLWAESEGTVACLGAEDRAATSAKSWPIFLFFAVILGGPYLIWKLLSTHSDEVTDSINWASGEDDHVVARAEYDFAAVSEEEISFRAGDMLNLALKEQQPKVRGWLLASLDGQTTGLIPANYVKILGKRKGRKTVESSKVSKQQQSFTNPTLTKGATVADSLDEQEAAFESVFVETNKVPVAPDSIGKDGEKQDL
Especificaciones

Especificaciones

NP_002609.1
Liquid
5194
PEX13 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASQPPPPPKPWETRRIPGAGPGPGPGPTFQSADLGPTLMTRPGQPALTRVPPPILPRPSQQTGSSSVNTFRPAYSSFSSGYGAYGNSFYGGYSPYSYGYNGLGYNRLRVDDLPPSRFVQQAEESSRGAFQSIESIVHAFASVSMMMDATFSAVYNSFRAVLDVANHFSRLKIHFTKVFSAFALVRTIRYLYRRLQRMLGLRRGSENEDLWAESEGTVACLGAEDRAATSAKSWPIFLFFAVILGGPYLIWKLLSTHSDEVTDSINWASGEDDHVVARAEYDFAAVSEEEISFRAGDMLNLALKEQQPKVRGWLLASLDGQTTGLIPANYVKILGKRKGRKTVESSKVSKQQQSFTNPTLTKGATVADSLDEQEAAFESVFVETNKVPVAPDSIGKDGEKQDL
RUO
PEX13
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
NALD/ZWS
PEX13
Yes
wheat germ expression system
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales