missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PDPN Full-length ORF (AAH22812.2, 1 a.a. - 238 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00010630-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16105462

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a type-I integral membrane glycoprotein with diverse distribution in human tissues. The physiological function of this protein may be related to its mucin-type character. The homologous protein in other species has been described as a differentiation antigen and influenza-virus receptor. The specific function of this protein has not been determined but it has been proposed as a marker of lung injury. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MLTPLGKFSTAKFAVRLPRVWEARAPSLSGAPAPTPPAPPPSRSSRLGLWPRCFLIFPQLRILLLGPQESNNSTGTMWKVSALLFVLGSASLWVLAEGASTGQPEDDTETTGLEGGVAMPGAEDDVVTPGTSEDRYKSGLTTLVATSVNSVTGIRIEDLPTSESTVHAQEQSPSATASNVATSHSTEKVDGDTQTTVEKDGLSTVTLVGIIVGVLLAIGFIGGIIVVVMRKMSGRYSP
Especificaciones

Especificaciones

AAH22812.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AGGRUS/GP36/GP40/Gp38/HT1A-1/OTS8/PA2.26/T1A/T1A-2
PDPN
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
10630
PDPN (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MLTPLGKFSTAKFAVRLPRVWEARAPSLSGAPAPTPPAPPPSRSSRLGLWPRCFLIFPQLRILLLGPQESNNSTGTMWKVSALLFVLGSASLWVLAEGASTGQPEDDTETTGLEGGVAMPGAEDDVVTPGTSEDRYKSGLTTLVATSVNSVTGIRIEDLPTSESTVHAQEQSPSATASNVATSHSTEKVDGDTQTTVEKDGLSTVTLVGIIVGVLLAIGFIGGIIVVVMRKMSGRYSP
RUO
PDPN
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales