Learn More
Abnova™ Human PCSK1N Partial ORF (NP_037403.1, 173 a.a. - 260 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00027344-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Members of the subtilisin-like proprotein convertase family process latent precursor proteins into their biologically active products. The protein encoded by this gene appears to function as an endogenous inhibitor of proprotein convertase subtilisin/kexin type 1. [provided by RefSeq]
Sequence: DGPAGPDAEEAGDETPDVDPELLRYLLGRILAGSADSEGVAAPRRLRRAADHDVGSELPPEGVLGALLRVKRLETPAPQVPARRLLPPEspecificaciones
NP_037403.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.42kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DGPAGPDAEEAGDETPDVDPELLRYLLGRILAGSADSEGVAAPRRLRRAADHDVGSELPPEGVLGALLRVKRLETPAPQVPARRLLPP | |
RUO | |
PCSK1N | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
27344 | |
PCSK1N (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PROSAAS/SAAS | |
PCSK1N | |
Recombinant | |
wheat germ expression system |