missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCGF6 Full-length ORF (AAH07602.1, 1 a.a. - 277 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00084108-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16195633

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The protein encoded by this gene contains a RING finger motif, which is most closely related to those of polycomb group (PcG) proteins RNF110/MEL-18 and BMI1. PcG proteins are known to form protein complexes and function as transcription repressors. This protein has been shown to interact with some PcG proteins and act as a transcription repressor. The activity of this protein is found to be regulated by cell cycle dependent phosphorylation. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MEGVAVVTAGSVGAAKTEGAAALPPPPPPPVSPPALTPAPAAGEEGPAPLSETGAPGCSGSRPPELEPERSLGRFRGRFEDEDEELEEEEELEEEEEEEEEDMSHFSLRLEGGRQDSEDEEERLINLSELTPYILCSICKGYLIDATTITECLHTFCKSCIVRHFYYSNRCPKCNIVVHQTQPLYNISANEGTGHFKPLEKKFVRVSGEATIGHVEKFLRRKMGLDPACQVDIICGDHLLEQYQTLREIRRAIGDAAMQDGLLVLHYGLVVSPLKIT
Especificaciones

Especificaciones

AAH07602.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
56.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MBLR/MGC15678/MGC17541/RNF134
PCGF6
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84108
PCGF6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEGVAVVTAGSVGAAKTEGAAALPPPPPPPVSPPALTPAPAAGEEGPAPLSETGAPGCSGSRPPELEPERSLGRFRGRFEDEDEELEEEEELEEEEEEEEEDMSHFSLRLEGGRQDSEDEEERLINLSELTPYILCSICKGYLIDATTITECLHTFCKSCIVRHFYYSNRCPKCNIVVHQTQPLYNISANEGTGHFKPLEKKFVRVSGEATIGHVEKFLRRKMGLDPACQVDIICGDHLLEQYQTLREIRRAIGDAAMQDGLLVLHYGLVVSPLKIT
RUO
PCGF6
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales