missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PCGF2 Partial ORF (AAH04858.1, 236 a.a. - 294 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

341.70€ - 518.16€

Especificaciones

Nombre PCGF2 (Human) Recombinant Protein (Q02)
Recombinante Yes
Para utilizar con (aplicación) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Peso molecular 32.23kDa
Requisitos de almacenamiento Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
16197835
Ver documentos Detalles de la promoción
Abnova™
H00007703-Q02.10UG
10 ug
341.70€
10 microgramos
Ver productos alternativos
Ahorre hasta 
»
Envío estimado: 14-03-2022
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16107845
Ver documentos Detalles de la promoción
Abnova™
H00007703-Q02.25UG
25 ug
518.16€
25 microgramos
Ver productos alternativos
Ahorre hasta 
»
Envío estimado: 14-03-2022
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

The protein encoded by this gene contains a RING finger motif and is similar to the polycomb group (PcG) gene products. PcG gene products form complexes via protein-protein interaction and maintain the transcription repression of genes involved in embryogenesis, cell cycles, and tumorigenesis. This protein was shown to act as a negative regulator of transcription and has tumor suppressor activity. The expression of this gene was detected in various tumor cells, but is limited in neural organs in normal tissues. Knockout studies in mice suggested that this protein may negatively regulate the expression of different cytokines, chemokines, and chemokine receptors, and thus plays an important role in lymphocyte differentiation and migration, as well as in immune responses. [provided by RefSeq]

Sequence: LTLATVPTPSEGTNTSGASECESVSDKAPSPATLPATSSSLPSPATPSHGSPSSHGPPA
Especificaciones

Especificaciones

PCGF2 (Human) Recombinant Protein (Q02)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
32.23kDa
SDS
Product Certifications