Learn More
Abnova™ Human PATZ1 Partial ORF (NP_114440.1, 260 a.a. - 358 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Especificaciones
Número de acceso | NP_114440.1 |
---|---|
Para utilizar con (aplicación) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID de gen (Entrez) | 23598 |
Peso molecular | 36.63kDa |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16194886
|
Abnova™
H00023598-Q01.25UG |
25 ug |
508.00€
25 microgramos |
Fecha estimada de envío: 05-06-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
16184886
|
Abnova™
H00023598-Q01.10UG |
10 ug |
335.00€
10 microgramos |
Fecha estimada de envío: 05-06-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
The protein encoded by this gene contains an A-T hook DNA binding motif which usually binds to other DNA binding structures to play an important role in chromatin modeling and transcription regulation. Its Poz domain is thought to function as a site for protein-protein interaction and is required for transcriptional repression, and the zinc-fingers comprise the DNA binding domain. Since the encoded protein has typical features of a transcription factor, it is postulated to be a repressor of gene expression. In small round cell sarcoma, this gene is fused to EWS by a small inversion of 22q, then the hybrid is thought to be translocated (t(1;22)(p36.1;q12). The rearrangement of chromosome 22 involves intron 8 of EWS and exon 1 of this gene creating a chimeric sequence containing the transactivation domain of EWS fused to zinc finger domain of this protein. This is a distinct example of an intra-chromosomal rearrangement of chromosome 22. Four alternatively spliced transcript variants are described for this gene. [provided by RefSeq]
Sequence: PLTGKRGRGRPRKANLLDSMFGSPGGLREAGILPCGLCGKVFTDANRLRQHEAQHGVTSLQLGYIDLPPPRLGENGLPISEDPDGPRKRSRTRKQVACEEspecificaciones
NP_114440.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAZR/PATZ/RIAZ/ZBTB19/ZNF278/ZSG/dJ400N23 | |
PATZ1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23598 | |
PATZ1 (Human) Recombinant Protein (Q01) | |
PLTGKRGRGRPRKANLLDSMFGSPGGLREAGILPCGLCGKVFTDANRLRQHEAQHGVTSLQLGYIDLPPPRLGENGLPISEDPDGPRKRSRTRKQVACE | |
RUO | |
PATZ1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |