Learn More
Abnova™ Human PARP4 Partial ORF (NP_006428, 31 a.a. - 140 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00000143-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes poly(ADP-ribosyl)transferase-like 1 protein, which is capable of catalyzing a poly(ADP-ribosyl)ation reaction. This protein has a catalytic domain which is homologous to that of poly (ADP-ribosyl) transferase, but lacks an N-terminal DNA binding domain which activates the C-terminal catalytic domain of poly (ADP-ribosyl) transferase. Since this protein is not capable of binding DNA directly, its transferase activity may be activated by other factors such as protein-protein interaction mediated by the extensive carboxyl terminus. [provided by RefSeq]
Sequence: KENGGKFSFSLNPQCTHIILDNADVLSQYQLNSIQKNHVHIANPDFIWKSIREKRLLDVKNYDPYKPLDITPPPDQKASSSEVKTEGLCPDSATEEEDTVELTEFGMQNVEspecificaciones
NP_006428 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KENGGKFSFSLNPQCTHIILDNADVLSQYQLNSIQKNHVHIANPDFIWKSIREKRLLDVKNYDPYKPLDITPPPDQKASSSEVKTEGLCPDSATEEEDTVELTEFGMQNV | |
RUO | |
PARP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
143 | |
PARP4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ADPRTL1/PARPL/PH5P/VAULT3/VPARP/VWA5C/p193 | |
PARP4 | |
Recombinant | |
wheat germ expression system |