missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PARP15 Full-length ORF (NP_689828.1, 1 a.a. - 444 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00165631-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16109023

  • 518.16€ / 25 microgramos
Envío estimado: 20-01-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

PARP15 is a macrodomain-containing transcriptional repressor with poly(ADP-ribose) polymerase activity (Aguiar et al., 2005 [PubMed 16061477]).[supplied by OMIM]

Sequence: MLQRIGLIFLHNIVVVSNCFYFQAFLDEFTNWSRINPNKARIPMAGDTQGVVGTVSKPCFTAYEMKIGAITFQVATGDIATEQVDVIVNSTARTFNRKSGVSRAILEGAGQAVESECAVLAAQPHRDFIITPGGCLKCKIIIHVPGGKDVRKTVTSVLEECEQRKYTSVSLPAIGTGNAGKNPITVADNIIDAIVDFSSQHSTPSLKTVKVVIFQPELLNIFYDSMKKRDLSASLNFQSTFSMTTCNLPEHWTDMNHQLFCMVQLEPGQSEYNTIKDKFTRTCSSYAIEKIERIQNAFLWQSYQVKKRQMDIKNDHKNNERLLFHGTDADSVPYVNQHGFNRSCAGKNAVSYGKGTYFAVDASYSAKDTYSKPDSNGRKHMYVVRVLTGVFTKGRAGLVTPPPKNPHNPTDLFDSVTNNTRSPKLFVVFFDNQAYPEYLITFTA
Especificaciones

Especificaciones

NP_689828.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BAL3/FLJ40196/FLJ40597/MGC126750/MGC126752
PARP15
Yes
wheat germ expression system
Liquid
165631
PARP15 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLQRIGLIFLHNIVVVSNCFYFQAFLDEFTNWSRINPNKARIPMAGDTQGVVGTVSKPCFTAYEMKIGAITFQVATGDIATEQVDVIVNSTARTFNRKSGVSRAILEGAGQAVESECAVLAAQPHRDFIITPGGCLKCKIIIHVPGGKDVRKTVTSVLEECEQRKYTSVSLPAIGTGNAGKNPITVADNIIDAIVDFSSQHSTPSLKTVKVVIFQPELLNIFYDSMKKRDLSASLNFQSTFSMTTCNLPEHWTDMNHQLFCMVQLEPGQSEYNTIKDKFTRTCSSYAIEKIERIQNAFLWQSYQVKKRQMDIKNDHKNNERLLFHGTDADSVPYVNQHGFNRSCAGKNAVSYGKGTYFAVDASYSAKDTYSKPDSNGRKHMYVVRVLTGVFTKGRAGLVTPPPKNPHNPTDLFDSVTNNTRSPKLFVVFFDNQAYPEYLITFTA
RUO
PARP15
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales