missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PALM Partial ORF (NP_005839, 176 a.a. - 284 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005064-Q01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16102365

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 10-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene encodes a member of the paralemmin protein family. The product of this gene is a prenylated and palmitoylated phosphoprotein that associates with the cytoplasmic face of plasma membranes and is implicated in plasma membrane dynamics in neurons and other cell types. Several alternatively spliced transcript variants have been identified, but the full-length nature of only two transcript variants has been determined. [provided by RefSeq]

Sequence: VEKDKVTGETRVLSSTTLLPRQPLPLGIKVYEDETKVVHAVDGTAENGIHPLSSSEVDELIHKADEVTLSEAGSTAGAAETRGAVEGAARTTPSRREITGVQAQPGEAT
Especificaciones

Especificaciones

NP_005839
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VEKDKVTGETRVLSSTTLLPRQPLPLGIKVYEDETKVVHAVDGTAENGIHPLSSSEVDELIHKADEVTLSEAGSTAGAAETRGAVEGAARTTPSRREITGVQAQPGEAT
RUO
PALM
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
5064
PALM (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0270
PALM
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales