Learn More
Novus Biologicals™ Human OXTR ELISA Kit (Colorimetric)
This kit recognizes Human OTR in samples.
Marca:  Novus Biologicals™ NBP2-80262
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214096
  • 769.96€ / 1 set
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
No significant cross-reactivity or interference between Human OTR and analogues was observed.
Especificaciones
Especificaciones
P30559, P30559
0.094 ng/mL
Other Biological Fluids, Plasma, Serum
5021
Human
Cancer, GPCR, Neuroscience, Neurotransmission
OXTR
0.156 to 10 ng/mL
ELISA Kit (Colorimetric)
ELISA
OXTR
1 Kit
Storage of components varies. See protocol for specific instructions.