missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ORF1-FL49 Full-length ORF (AAH13643, 1 a.a. - 97 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00084418-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: MNQENPPPYPGPGPTAPYPPYPPQPMGPGPMGGPYPPPQGYPYQGYPQYGWQGGPQEPPKTTVYVVEDQRRDELGPSTCLTACWTALCCCCLWDMLTEspecificaciones
AAH13643 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.41kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ORF1-FL49 | |
C5orf32 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84418 | |
ORF1-FL49 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNQENPPPYPGPGPTAPYPPYPPQPMGPGPMGGPYPPPQGYPYQGYPQYGWQGGPQEPPKTTVYVVEDQRRDELGPSTCLTACWTALCCCCLWDMLT | |
RUO | |
C5orf32 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |