missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ORC2L Full-length ORF (AAH14834, 1 a.a. - 577 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004999-P01.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16108611

  • 341.70€ / 10 microgramos
Envío estimado: 20-01-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The origin recognition complex (ORC) is a highly conserved six subunits protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. This protein forms a core complex with ORC3L, -4L, and -5L. It also interacts with CDC45L and MCM10, which are proteins known to be important for the initiation of DNA replication. This protein has been demonstrated to specifically associate with the origin of replication of Epstein-Barr virus in human cells, and is thought to be required for DNA replication from viral origin of replication. [provided by RefSeq]

Sequence: MSKPELKEDKMLEVHFVGDDDVLNHILDREGGAKLKKERAQLLVNPKKIIKKPEYDLEEDDQEVLKDQNYVEIMGRDVQESLKNGSATGGGNKVYSFQNRKHSEKMAKLASELAKTPQKSVSFSLKNDPEITINVPQSSKGHSASDKVQPKNNDKSEFLSTAPRSLRKRLIVPRSHSDSESEYSASNSEDDEGVAQEHEEDTNAVIFSQKIQAQNRVVSAPVGKETPSKRMKRDKTSDLVEEYFEAHSSSKVLTSDRTLQKLKRAKLDQQTLRNLLSKVSPSFSAELKQLNQQYEKLFHKWMLQLHLGFNIVLYGLGSKRDLLERFRTTMLQDSIHVVINGFFPGISVKSVLNSITEEVLDHMGTFRSILDQLDWIVNKFKEDSSLELFLLIHNLDSQMLRGEKSQQIIGQLSSLHNIYLIASIDHLNAPLMWDHAKQSLFNWLWYETTTYSPYTEETSYENSLLVKQSGSLPLSSLTHVLRSLTPNARGIFRLLIKYQLDNQDNPSYIGLSFQDFYQQCREAFLVNSDLTLRAQLTEFRDHKLIRTKKGTDGVEYLLIPVDNGTLTDFLEKEEEEA
Especificaciones

Especificaciones

AAH14834
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
89.21kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ORC2
ORC2L
Yes
wheat germ expression system
Liquid
4999
ORC2L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSKPELKEDKMLEVHFVGDDDVLNHILDREGGAKLKKERAQLLVNPKKIIKKPEYDLEEDDQEVLKDQNYVEIMGRDVQESLKNGSATGGGNKVYSFQNRKHSEKMAKLASELAKTPQKSVSFSLKNDPEITINVPQSSKGHSASDKVQPKNNDKSEFLSTAPRSLRKRLIVPRSHSDSESEYSASNSEDDEGVAQEHEEDTNAVIFSQKIQAQNRVVSAPVGKETPSKRMKRDKTSDLVEEYFEAHSSSKVLTSDRTLQKLKRAKLDQQTLRNLLSKVSPSFSAELKQLNQQYEKLFHKWMLQLHLGFNIVLYGLGSKRDLLERFRTTMLQDSIHVVINGFFPGISVKSVLNSITEEVLDHMGTFRSILDQLDWIVNKFKEDSSLELFLLIHNLDSQMLRGEKSQQIIGQLSSLHNIYLIASIDHLNAPLMWDHAKQSLFNWLWYETTTYSPYTEETSYENSLLVKQSGSLPLSSLTHVLRSLTPNARGIFRLLIKYQLDNQDNPSYIGLSFQDFYQQCREAFLVNSDLTLRAQLTEFRDHKLIRTKKGTDGVEYLLIPVDNGTLTDFLEKEEEEA
RUO
ORC2L
Wheat Germ (in vitro)
GST
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales