missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OMP Full-length ORF (NP_006180.1, 1 a.a. - 163 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004975-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16108581

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 07-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Olfactory marker protein is uniquely associated with the mature olfactory receptor neurons in many vertebrate species from fish to man. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Results of the mouse knockout studies show that OMP-null mice are compromised in their ability to respond to odor stimuli, and that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. [provided by RefSeq]

Sequence: MAEDRPQQPQLDMPLVLDQGLTRQMRLRVESLKQRGEKRQDGEKLLQPAESVYRLNFTQQQRLQFERWNVVLDKPGKVTITGTSQNWTPDLTNLMTRQLLDPTAIFWRKEDSDAIDWNEADALEFGERLSDLAKIRKVMYFLVTFGEGVEPANLKASVVFNQL
Especificaciones

Especificaciones

NP_006180.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.3kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
OMP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
4975
OMP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAEDRPQQPQLDMPLVLDQGLTRQMRLRVESLKQRGEKRQDGEKLLQPAESVYRLNFTQQQRLQFERWNVVLDKPGKVTITGTSQNWTPDLTNLMTRQLLDPTAIFWRKEDSDAIDWNEADALEFGERLSDLAKIRKVMYFLVTFGEGVEPANLKASVVFNQL
RUO
OMP
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales