missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OFD1 Full-length ORF (AAH99658.1, 1 a.a. - 550 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008481-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16194918

  • 508.00€ / 25 microgramos
Fecha estimada de envío: 05-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

This gene is located on the X chromosome and encodes a centrosomal protein. A knockout mouse model has been used to study the effect of mutations in this gene. The mouse gene is also located on the X chromosome, however, unlike the human gene it is not subject to X inactivation. Mutations in this gene are associated with oral-facial-digital syndrome type I and Simpson-Golabi-Behmel syndrome type 2. Many pseudogenes have been identified; a single pseudogene is found on chromosome 5 while as many as fifteen have been found on the Y chromosome. Alternatively spliced transcripts have been described for this gene but the biological validity of these transcripts has not been determined. [provided by RefSeq]

Sequence: MLNEKVKEMSDYSLLKEEKLELLAQNKLLKQQLEESRNENLRLLNRLAQPAPELAVFQKELRKAEKAIVVEHEEFESCRQALHKQLQDEIEHSAQLKAQILGYKASVKSLTTQVADLKLQLKQTQTALENEVYCNPKQSVIDRSVNGLINGNVVPCNGEISGDFLNNPFKQENVLARMVASRITNYPTAWVEGSSPDSDLEFVANTKARVKELQQEAERLEKAFRSYHRRVIKNSAKSPLAAKSPPSLHLLEAFKNITSSSPERHIFGEDRVVSEQPQVGTLEERNDVVEALTGSAASRLRGGTSSRRLSSTPLPKAKRSLESEMYLEGLGRSHIASPSPCPDRMPLPSPTESRHSLSIPPVSSPPEQKVGLYRRQTELQDKSEFSDVDKLAFKDNEEFESSFECVDQKQIEEQKEEEKIREQQVKERRQREERRQSNLQEVLERERRELEKLYQERKMIEESLKIKIKKELEMENELEMSNQEIKDKSAHSENPLEKYMKIIQQEQDQESADKSSKKMVQEGSLVDTLQSSDKVESLTGFSHEELDDSW
Especificaciones

Especificaciones

AAH99658.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
89.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
71-7A/CXorf5/MGC117039/MGC117040/SGBS2
OFD1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8481
OFD1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLNEKVKEMSDYSLLKEEKLELLAQNKLLKQQLEESRNENLRLLNRLAQPAPELAVFQKELRKAEKAIVVEHEEFESCRQALHKQLQDEIEHSAQLKAQILGYKASVKSLTTQVADLKLQLKQTQTALENEVYCNPKQSVIDRSVNGLINGNVVPCNGEISGDFLNNPFKQENVLARMVASRITNYPTAWVEGSSPDSDLEFVANTKARVKELQQEAERLEKAFRSYHRRVIKNSAKSPLAAKSPPSLHLLEAFKNITSSSPERHIFGEDRVVSEQPQVGTLEERNDVVEALTGSAASRLRGGTSSRRLSSTPLPKAKRSLESEMYLEGLGRSHIASPSPCPDRMPLPSPTESRHSLSIPPVSSPPEQKVGLYRRQTELQDKSEFSDVDKLAFKDNEEFESSFECVDQKQIEEQKEEEKIREQQVKERRQREERRQSNLQEVLERERRELEKLYQERKMIEESLKIKIKKELEMENELEMSNQEIKDKSAHSENPLEKYMKIIQQEQDQESADKSSKKMVQEGSLVDTLQSSDKVESLTGFSHEELDDSW
RUO
OFD1
Wheat Germ (in vitro)
GST
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales