missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human OBSCN Partial ORF (XP_290923, 1551 a.a. - 1649 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00084033-Q02.10ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16100617

  • 335.00€ / 10 microgramos
Fecha estimada de envío: 05-06-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

The obscurin gene spans more than 150 kb, contains over 80 exons and encodes a protein of approximately 720 kDa. The encoded protein contains 68 Ig domains, 2 fibronectin domains, 1 calcium/calmodulin-binding domain, 1 RhoGEF domain with an associated PH domain, and 2 serine-threonine kinase domains. This protein belongs to the family of giant sacromeric signaling proteins that includes titin and nebulin, and may have a role in the organization of myofibrils during assembly and may mediate interactions between the sarcoplasmic reticulum and myofibrils. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: KAGMGPYSSPSEQVLLGGPSHLASEEESQGRSAQPLPSTKTFAFQTQIQRGRFSVVRQCWEKASGRALAAKIIPYHPKDKTAVLREYEALKGLRHPHLA
Especificaciones

Especificaciones

XP_290923
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KAGMGPYSSPSEQVLLGGPSHLASEEESQGRSAQPLPSTKTFAFQTQIQRGRFSVVRQCWEKASGRALAAKIIPYHPKDKTAVLREYEALKGLRHPHLA
RUO
OBSCN
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
84033
OBSCN (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp666E245/FLJ14124/KIAA1556/KIAA1639/MGC120409/MGC120410/MGC120411/MGC120412/MGC138590/UNC89
OBSCN
Recombinant
wheat germ expression system
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales