Learn More
Abnova™ Human NRK Partial ORF (NP_940867, 1483 a.a. - 1582 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00203447-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
NRK is a member of the GCK (MIM 138079) subfamily of protein kinases that are involved in activating the JNK pathway (Kanai-Azuma et al., 1999 [PubMed 10559491]).[supplied by OMIM]
Sequence: VEANEQLFKKILEMWKDIPSSIAFECTQRTTGWGQKAIEVRSLQSRVLESELKRRSIKKLRFLCTRGDKLFFTSTLRNHHSRVYFMTLGKLEELQSNYDVEspecificaciones
NP_940867 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VEANEQLFKKILEMWKDIPSSIAFECTQRTTGWGQKAIEVRSLQSRVLESELKRRSIKKLRFLCTRGDKLFFTSTLRNHHSRVYFMTLGKLEELQSNYDV | |
RUO | |
NRK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
203447 | |
NRK (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686A17109/FLJ16788/MGC131849/NESK | |
NRK | |
Recombinant | |
wheat germ expression system |