Learn More
Abnova™ Human NRIP1 Partial ORF (NP_003480, 1051 a.a. - 1158 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008204-Q01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Nuclear receptor interacting protein 1 (NRIP1) is a nuclear protein that specifically interacts with the hormone-dependent activation domain AF2 of nuclear receptors. Also known as RIP140, this protein modulates transcriptional activity of the estrogen receptor. [provided by RefSeq]
Sequence: KNEYEKDSPRLTKTNPILYYMLQKGGNSVTSRETQDKDIWREASSAESVSQVTAKEELLPTAETKASFFNLRSPYNSHMGNNASRPHSANGEVYGLLGSVLTIKKESEEspecificaciones
NP_003480 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KNEYEKDSPRLTKTNPILYYMLQKGGNSVTSRETQDKDIWREASSAESVSQVTAKEELLPTAETKASFFNLRSPYNSHMGNNASRPHSANGEVYGLLGSVLTIKKESE | |
RUO | |
NRIP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8204 | |
NRIP1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ77253/RIP140 | |
NRIP1 | |
Recombinant | |
wheat germ expression system |