Learn More
Abnova™ Human NPL Full-length ORF (AAH58003.1, 1 a.a. - 240 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00080896-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
N-acetylneuraminate pyruvate lyase (EC 4.1.3.3) controls the cellular concentration of sialic acid by catalyzing the conversion of sialic acid into acylmannosamines and pyruvate (Wu et al., 2005 [PubMed 16147865]).[supplied by OMIM]
Sequence: MAFPKKKLQGLVAATITPMTENGEINFSVIGQYVDYLVKEQGVKNIFVNGTTGEGLSLSVSERRQVAEEWVTKGKDKLDQVIIHVGALSLKESQELAQHAAEIGADGIAVIAPFFLKPWTKDILINFLKEVAAAAPALPFYYYHIPALTGVKIRAEELLDGILDKIPTFQGLKFSDTDLLDFGQCVDQNRQQQFAFLFGVDEFCIQRFINFVVKLENSKLKVSKNQRTLPLGTTNFPFLHEspecificaciones
AAH58003.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
53.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C1orf13/MGC149582/MGC61869/NPL1/c112 | |
NPL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
80896 | |
NPL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAFPKKKLQGLVAATITPMTENGEINFSVIGQYVDYLVKEQGVKNIFVNGTTGEGLSLSVSERRQVAEEWVTKGKDKLDQVIIHVGALSLKESQELAQHAAEIGADGIAVIAPFFLKPWTKDILINFLKEVAAAAPALPFYYYHIPALTGVKIRAEELLDGILDKIPTFQGLKFSDTDLLDFGQCVDQNRQQQFAFLFGVDEFCIQRFINFVVKLENSKLKVSKNQRTLPLGTTNFPFLH | |
RUO | |
NPL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |