Learn More
Abnova™ Human NPHP3 Partial ORF (NP_694972.3, 106 a.a. - 205 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00027031-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein containing a coiled-coil (CC) domain, a tubulin-tyrosine ligase (TTL) domain, and a tetratrico peptide repeat (TPR) domain. The encoded protein interacts with nephrocystin and may function in renal tubular development and function. Mutations in this gene are associated with nephronophthisis type 3. Multiple splice variants have been described but their full-length nature has not been determined. [provided by RefSeq]
Sequence: NQELLSMGRREAKLDTENKRLRAELQALQKTYQKILREKESALEAKYQAMERAATFEHDRDKVKRQFKIFRETKENEIQDLLRAKRELESKLQRLQAQGIEspecificaciones
NP_694972.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NQELLSMGRREAKLDTENKRLRAELQALQKTYQKILREKESALEAKYQAMERAATFEHDRDKVKRQFKIFRETKENEIQDLLRAKRELESKLQRLQAQGI | |
RUO | |
NPHP3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
27031 | |
NPHP3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp667K242/DKFZp781K1312/FLJ30691/FLJ36696/KIAA2000/MGC78666/NPH3 | |
NPHP3 | |
Recombinant | |
wheat germ expression system |