Learn More
Abnova™ Human NOXO1 Partial ORF (AAH15917, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00124056-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
NADPH oxidases (NOXs) catalyze the transfer of electrons from NADPH to molecular oxygen to generate reactive oxygen species (ROS). NOX organizers, such as NOXO1, target NOX activators (see NOXA1; MIM 611255) to NOX and also target NOX to different subcellular compartments (Opitz et al., 2007 [PubMed 17189823]).[supplied by OMIM]
Sequence: MAGPRYPVSVQGAALVQIKRLQTFAFSVRWSDGSDTFVRRSWDEFRQLKTLKETFPVEAGLLRRSDRVLPKLLDAPLLGRVGRTSRGLARLQLLETYSRREspecificaciones
AAH15917 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAGPRYPVSVQGAALVQIKRLQTFAFSVRWSDGSDTFVRRSWDEFRQLKTLKETFPVEAGLLRRSDRVLPKLLDAPLLGRVGRTSRGLARLQLLETYSRR | |
RUO | |
NOXO1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
124056 | |
NOXO1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC20258/P41NOX/P41NOXA/P41NOXB/P41NOXC/SH3PXD5/SNX28 | |
NOXO1 | |
Recombinant | |
wheat germ expression system |