missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NLN Full-length ORF (NP_065777.1, 1 a.a. - 704 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00057486-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Neurolysin (EC 3.4.24.16) is an enzyme involved in the metabolic inactivation of bioactive peptides (Garrido et al., 1999 [PubMed 10235115]).[supplied by OMIM]
Sequence: MIARCLLAVRSLRRVGGSRILLRMTLGREVMSPLQAMSSYTVAGRNVLRWDLSPEQIKTRTEELIVQTKQVYDAVGMLGIEEVTYENCLQALADVEVKYIVERTMLDFPQHVSSDKEVRAASTEADKRLSRFDIEMSMRGDIFERIVHLQETCDLGKIKPEARRYLEKSIKMGKRNGLHLPEQVQNEIKSMKKRMSELCIDFNKNLNEDDTFLVFSKAELGALPDDFIDSLEKTDDDKYKITLKYPHYFPVMKKCCIPETRRRMEMAFNTRCKEENTIILQQLLPLRTKVAKLLGYSTHADFVLEMNTAKSTSRVTAFLDDLSQKLKPLGEAEREFILNLKKKECKDRGFEYDGKINAWDLYYYMTQTEELKYSIDQEFLKEYFPIEVVTEGLLNTYQELLGLSFEQMTDAHVWNKSVTLYTVKDKATGEVLGQFYLDLYPREGKYNHAACFGLQPGCLLPDGSRMMAVAALVVNFSQPVAGRPSLLRHDEVRTYFHEFGHVMHQICAQTDFARFSGTNVETDFVEVPSQMLENWVWDVDSLRRLSKHYKDGSPIADDLLEKLVASRLVNTGLLTLRQIVLSKVDQSLHTNTSLDAASEYAKYCSEILGVAATPGTNMPATFGHLAGGYDGQYYGYLWSEVFSMDMFYSCFKKEGIMNPEVGMKYRNLILKPGGSLDGMDMLHNFLKREPNQKAFLMSRGLHAPEspecificaciones
NP_065777.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
107.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AGTBP/DKFZp564F123/KIAA1226 | |
NLN | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
57486 | |
NLN (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIARCLLAVRSLRRVGGSRILLRMTLGREVMSPLQAMSSYTVAGRNVLRWDLSPEQIKTRTEELIVQTKQVYDAVGMLGIEEVTYENCLQALADVEVKYIVERTMLDFPQHVSSDKEVRAASTEADKRLSRFDIEMSMRGDIFERIVHLQETCDLGKIKPEARRYLEKSIKMGKRNGLHLPEQVQNEIKSMKKRMSELCIDFNKNLNEDDTFLVFSKAELGALPDDFIDSLEKTDDDKYKITLKYPHYFPVMKKCCIPETRRRMEMAFNTRCKEENTIILQQLLPLRTKVAKLLGYSTHADFVLEMNTAKSTSRVTAFLDDLSQKLKPLGEAEREFILNLKKKECKDRGFEYDGKINAWDLYYYMTQTEELKYSIDQEFLKEYFPIEVVTEGLLNTYQELLGLSFEQMTDAHVWNKSVTLYTVKDKATGEVLGQFYLDLYPREGKYNHAACFGLQPGCLLPDGSRMMAVAALVVNFSQPVAGRPSLLRHDEVRTYFHEFGHVMHQICAQTDFARFSGTNVETDFVEVPSQMLENWVWDVDSLRRLSKHYKDGSPIADDLLEKLVASRLVNTGLLTLRQIVLSKVDQSLHTNTSLDAASEYAKYCSEILGVAATPGTNMPATFGHLAGGYDGQYYGYLWSEVFSMDMFYSCFKKEGIMNPEVGMKYRNLILKPGGSLDGMDMLHNFLKREPNQKAFLMSRGLHAP | |
RUO | |
NLN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |