Learn More
Abnova™ Human NINJ1 Full-length ORF (NP_004139.1, 1 a.a. - 152 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004814-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
The ninjurin protein is upregulated after nerve injury both in dorsal root ganglion neurons and in Schwann cells (Araki and Milbrandt, 1996 [PubMed 8780658]). It demonstrates properties of a homophilic adhesion molecule and promotes neurite outgrowth from primary cultured dorsal root ganglion neurons.[supplied by OMIM]
Sequence: MDSGTEEYELNGGLPPGTPGSPDASPARWGWRHGPINVNHYASKKSAAESMLDIALLMANASQLKAVVEQGPSFAFYVPLVVLISISLVLQIGVGVLLIFLVKYDLNNPDKHAKLDFLNNLATGLVFIIVVVNIFITAFGVQKPLMDMAPQQEspecificaciones
NP_004139.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
42.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NIN1/NINJURIN | |
NINJ1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4814 | |
NINJ1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDSGTEEYELNGGLPPGTPGSPDASPARWGWRHGPINVNHYASKKSAAESMLDIALLMANASQLKAVVEQGPSFAFYVPLVVLISISLVLQIGVGVLLIFLVKYDLNNPDKHAKLDFLNNLATGLVFIIVVVNIFITAFGVQKPLMDMAPQQ | |
RUO | |
NINJ1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |