missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human NHS Partial ORF (NP_938011.1, 1532 a.a. - 1629 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00004810-Q01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
Sequence: TAESPQSTDDAHQGSQGAEALSPLSPCSPRVNAEGFSSKSFATSASARVGRSRAPPAASSSRYSVRCRLYNTPMQAISEGETENSDGSPHDDRSSQSSEspecificaciones
NP_938011.1 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TAESPQSTDDAHQGSQGAEALSPLSPCSPRVNAEGFSSKSFATSASARVGRSRAPPAASSSRYSVRCRLYNTPMQAISEGETENSDGSPHDDRSSQSS | |
RUO | |
NHS | |
Wheat Germ (in vitro) | |
GST |
wheat germ expression system | |
Liquid | |
4810 | |
NHS (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp781F2016|DKFZp781L0254|SCML1 | |
NHS | |
Yes |